Learn More
Biomatik Corporation DNAJB8 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09538
Encompass_Preferred
The DNAJB8 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human DnaJ homolog subfamily B member 8 protein (1-232AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: DnaJ homolog subfamily B member 8, DNAJB8. UniProt: Q8NHS0. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9120