Learn More
Biomatik Corporation ASAP3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09539
Encompass_Preferred
The ASAP3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 3 protein (451-700AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 3, Development and differentiation-enhancing factor-like 1, Protein up-regulated in liver cancer 1, ASAP3, DDEFL1, UPLC1. UniProt: Q8TDY4. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9121