Learn More
Biomatik Corporation RPGR anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09543
Encompass_Preferred
Rpgr Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RPGR. The Rpgr Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human X-Linked Retinitis Pigmentosa Gtpase Regulator Protein (1-180Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: RPGR. Accession Number: Q92834. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9125