Learn More
Biomatik Corporation ARRDC3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09545
Encompass_Preferred
The ARRDC3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Arrestin domain-containing protein 3 protein (1-270AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-1:5000. Alternative Names: Arrestin domain-containing protein 3, ARRDC3, KIAA1376, TBP-2-like inducible membrane protein1. UniProt: Q96B67. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9127