Learn More
Biomatik Corporation PGAM5 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09548
Encompass_Preferred
Pgam5 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PGAM5. The Pgam5 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Serine; Threonine-Protein Phosphatase Pgam5, Mitochondrial Protein (30-223Aa). Applications: ELISA, IHC, IF. Target Synonyms: PGAM5. Accession Number: Q96HS1. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9130