Learn More
Biomatik Corporation SLC35F5 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09558
Encompass_Preferred
Slc35F5 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC35F5. The Slc35F5 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Solute Carrier Family 35 Member F5 Protein (122-242Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SLC35F5. Accession Number: Q8WV83. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9140