Learn More
Biomatik Corporation RQCD1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509141
Encompass_Preferred
Cnot9 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to CNOT9. The Cnot9 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ccr4-Not Transcription Complex Subunit 9 Protein (1-258Aa). Applications: ELISA, WB, IHC. Target Synonyms: CNOT9. Accession Number: Q92600. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9141 Supplier Biomatik Corporation Supplier No. CAC09559
Cnot9 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to CNOT9. The Cnot9 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ccr4-Not Transcription Complex Subunit 9 Protein (1-258Aa). Applications: ELISA, WB, IHC. Target Synonyms: CNOT9. Accession Number: Q92600. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.