Learn More
Biomatik Corporation YIPF5 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09562
Encompass_Preferred
Yipf5 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to YIPF5. The Yipf5 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Protein Yipf5 Protein (1-124Aa). Applications: ELISA, WB, IF. Target Synonyms: YIPF5. Accession Number: Q969M3. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9144