Learn More
Biomatik Corporation LRRC57 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09578
Encompass_Preferred
Lrrc57 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LRRC57. The Lrrc57 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Leucine-Rich Repeat-Containing Protein 57 Protein (1-239Aa). Applications: ELISA, WB. Target Synonyms: LRRC57. Accession Number: Q8N9N7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9160