Learn More
Biomatik Corporation CLEC1A anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09580
Encompass_Preferred
The CLEC1A Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human C-type lectin domain family 1 member A protein (74-280AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: C-type lectin domain family 1 member A, CLEC1A, CLEC1, UNQ569/PRO1131, C-type lectin-like receptor 1, CLEC-1. UniProt: Q8NC01. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9162