Learn More
Biomatik Corporation LMO3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09586
Encompass_Preferred
Lmo3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LMO3. The Lmo3 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Lim Domain Only Protein 3 Protein (1-145Aa). Applications: ELISA, WB, IHC. Target Synonyms: LMO3. Accession Number: Q8TAP4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9168