Learn More
Biomatik Corporation ATP6V0A1 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09593
Encompass_Preferred
The ATP6V0A1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant Human V-type proton ATPase 116 kDa subunit a isoform 1 protein (80-260AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: V-type proton ATPase 116 kDa subunit a isoform 1, V-ATPase 116 kDa isoform a1, Clathrin-coated vesicle/synaptic vesicle proton pump 116 kDa subunit, Vacuolar adenosine triphosphatase subunit Ac116, Vacuolar proton pump subunit 1, Vacuolar proton translocating ATPase 116 kDa subunit a isoform 1, ATP6V0A1, ATP6N1, ATP6N1A, VPP1. UniProt: Q93050. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9175