Learn More
Biomatik Corporation KLRG1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09600
Encompass_Preferred
Klrg1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to KLRG1. The Klrg1 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Killer Cell Lectin-Like Receptor Subfamily G Member 1 Protein (60-195Aa). Applications: ELISA, IHC. Target Synonyms: KLRG1. Accession Number: Q96E93. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9182