Learn More
Biomatik Corporation NEUROD6 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09602
Encompass_Preferred
Neurod6 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NEUROD6. The Neurod6 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Neurogenic Differentiation Factor 6 Protein (170-314Aa). Applications: ELISA, WB. Target Synonyms: NEUROD6. Accession Number: Q96NK8. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9184