Learn More
Biomatik Corporation TGS1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09607
Encompass_Preferred
Tgs1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TGS1. The Tgs1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Trimethylguanosine Synthase Protein (713-853Aa). Applications: ELISA, WB, IHC. Target Synonyms: TGS1. Accession Number: Q96RS0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9189