Learn More
Biomatik Corporation CA13 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09610
Encompass_Preferred
The CA13 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Carbonic anhydrase 13 protein (1-262AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Carbonic anhydrase 13, Carbonate dehydratase XIII, Carbonic anhydrase XIII, CA-XIII, CA13. UniProt: Q8N1Q1. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9192