Learn More
Biomatik Corporation TCP11L2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509194
Encompass_Preferred
Tcp11L2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TCP11L2. The Tcp11L2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human T-Complex Protein 11-Like Protein 2 Protein (1-210Aa). Applications: ELISA, WB, IHC. Target Synonyms: TCP11L2. Accession Number: Q8N4U5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9194 Supplier Biomatik Corporation Supplier No. CAC09612
Tcp11L2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TCP11L2. The Tcp11L2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human T-Complex Protein 11-Like Protein 2 Protein (1-210Aa). Applications: ELISA, WB, IHC. Target Synonyms: TCP11L2. Accession Number: Q8N4U5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.