Learn More
Biomatik Corporation TET1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09616
Encompass_Preferred
Tet1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TET1. The Tet1 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Methylcytosine Dioxygenase Tet1 Protein (1700-1800Aa). Applications: ELISA, IHC. Target Synonyms: TET1. Accession Number: Q8NFU7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9198