Learn More
Biomatik Corporation FBXO25 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509201
Encompass_Preferred
The FBXO25 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human F-box only protein 25 protein (1-291AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: F-box only protein 25, FBXO25, FBX25. UniProt: Q8TCJ0. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9201 Supplier Biomatik Corporation Supplier No. CAC09619
The FBXO25 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human F-box only protein 25 protein (1-291AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: F-box only protein 25, FBXO25, FBX25. UniProt: Q8TCJ0. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.