Learn More
Biomatik Corporation GCDH anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509206
Encompass_Preferred
The GCDH Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Glutaryl-CoA dehydrogenase, mitochondrial protein (45-300AA). Applications: ELISA, WB, IHC, IP. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IP:1:200-1:2000. Alternative Names: Glutaryl-CoA dehydrogenase, mitochondrial, GCD, GCDH. UniProt: Q92947. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9206 Supplier Biomatik Corporation Supplier No. CAC09624
The GCDH Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Glutaryl-CoA dehydrogenase, mitochondrial protein (45-300AA). Applications: ELISA, WB, IHC, IP. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IP:1:200-1:2000. Alternative Names: Glutaryl-CoA dehydrogenase, mitochondrial, GCD, GCDH. UniProt: Q92947. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.