Learn More
Biomatik Corporation RERG anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09626
Encompass_Preferred
Rerg Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RERG. The Rerg Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ras-Related And Estrogen-Regulated Growth Inhibitor Protein (1-199Aa). Applications: ELISA, WB, IHC. Target Synonyms: RERG. Accession Number: Q96A58. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9208