Learn More
Biomatik Corporation NSUN4 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509211
Encompass_Preferred
Nsun4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NSUN4. The Nsun4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 5-Methylcytosine Rrna Methyltransferase Nsun4 Protein (26-284Aa). Applications: ELISA, WB, IHC. Target Synonyms: NSUN4. Accession Number: Q96CB9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9211 Supplier Biomatik Corporation Supplier No. CAC09629
Nsun4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NSUN4. The Nsun4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 5-Methylcytosine Rrna Methyltransferase Nsun4 Protein (26-284Aa). Applications: ELISA, WB, IHC. Target Synonyms: NSUN4. Accession Number: Q96CB9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.