Learn More
Biomatik Corporation GPR146 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09630
Encompass_Preferred
The GPR146 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Probable G-protein coupled receptor 146 protein (301-333AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Probable G-protein coupled receptor 146, GPR146, PGR8, G-protein coupled receptor PGR8. UniProt: Q96CH1. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9212