Learn More
Biomatik Corporation APOPT1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509214
Encompass_Preferred
The APOPT1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Apoptogenic protein 1, mitochondrial protein (40-150AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-1:5000. Alternative Names: Apoptogenic protein 1, mitochondrial, APOP-1, APOPT1, APOP1, C14orf153. UniProt: Q96IL0. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9214 Supplier Biomatik Corporation Supplier No. CAC09632
The APOPT1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Apoptogenic protein 1, mitochondrial protein (40-150AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-1:5000. Alternative Names: Apoptogenic protein 1, mitochondrial, APOP-1, APOPT1, APOP1, C14orf153. UniProt: Q96IL0. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.