Learn More
Biomatik Corporation CD177 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509221
Encompass_Preferred
The CD177 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human CD177 antigen protein (22-321AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:2000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: CD177 antigen, Human neutrophil alloantigen 2a, HNA-2a, NB1 glycoprotein, NB1 GP, Polycythemia rubra vera protein 1, PRV-1, CD177, CD177, NB1, PRV1, UNQ595/PRO1181. UniProt: Q8N6Q3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9221 Supplier Biomatik Corporation Supplier No. CAC09639
The CD177 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human CD177 antigen protein (22-321AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:2000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: CD177 antigen, Human neutrophil alloantigen 2a, HNA-2a, NB1 glycoprotein, NB1 GP, Polycythemia rubra vera protein 1, PRV-1, CD177, CD177, NB1, PRV1, UNQ595/PRO1181. UniProt: Q8N6Q3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.