Learn More
Biomatik Corporation HM13 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09642
Encompass_Preferred
The HM13 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Minor histocompatibility antigen H13 protein (336-377AA). Applications: ELISA, WB, IF. Recommended dilution: WB:1:1000-1:5000, IF:1:50-1:200. Alternative Names: Minor histocompatibility antigen H13 (EC:3.4.23.-), HM13, H13, IMP1, PSL3, SPP, MSTP086, Intramembrane protease 1, IMP-1, IMPAS-1, hIMP1, Presenilin-like protein 3, Signal peptide peptidase. UniProt: Q8TCT9. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9224