Learn More
Biomatik Corporation OXR1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09664
Encompass_Preferred
Oxr1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to OXR1. The Oxr1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Oxidation Resistance Protein 1 Protein (401-600Aa). Applications: ELISA, WB, IHC. Target Synonyms: OXR1. Accession Number: Q8N573. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9246