Learn More
Biomatik Corporation KRT35 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09677
Encompass_Preferred
Krt35 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to KRT35. The Krt35 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Keratin, Type I Cuticular Ha5 Protein (1-455Aa). Applications: ELISA, WB, IHC. Target Synonyms: KRT35. Accession Number: Q92764. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9259