Learn More
Biomatik Corporation MYLPF anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509262
Encompass_Preferred
Mylpf Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MYLPF. The Mylpf Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Myosin Regulatory Light Chain 2, Skeletal Muscle Isoform Protein (2-169Aa). Applications: ELISA, WB. Target Synonyms: MYLPF. Accession Number: Q96A32. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9262 Supplier Biomatik Corporation Supplier No. CAC09680
Mylpf Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MYLPF. The Mylpf Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Myosin Regulatory Light Chain 2, Skeletal Muscle Isoform Protein (2-169Aa). Applications: ELISA, WB. Target Synonyms: MYLPF. Accession Number: Q96A32. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.