Learn More
Biomatik Corporation NOL4L anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09686
Encompass_Preferred
Nol4L Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NOL4L. The Nol4L Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Nucleolar Protein 4-Like Protein (101-300Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: NOL4L. Accession Number: Q96MY1. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9268