Learn More
Biomatik Corporation GPA33 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09689
Encompass_Preferred
The GPA33 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Cell surface A33 antigen protein (22-235AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-5000, IHC:1:20-1:200. Alternative Names: Cell surface A33 antigen, Glycoprotein A33, GPA33. UniProt: Q99795. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9271