Learn More
Biomatik Corporation EIF3C anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09690
Encompass_Preferred
The EIF3C Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Eukaryotic translation initiation factor 3 subunit C protein (201-500AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:2000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Eukaryotic translation initiation factor 3 subunit C, Eukaryotic translation initiation factor 3 subunit 8, EIF3C. UniProt: Q99613. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9272