Learn More
Biomatik Corporation FOXC2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09691
Encompass_Preferred
The FOXC2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Forkhead box protein C2 protein (24-294AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Forkhead box protein C2, Forkhead-related protein FKHL14, Mesenchyme fork head protein 1, MFH-1 protein, Transcription factor FKH-14, FOXC2, FKHL14, MFH1. UniProt: Q99958. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9273