Learn More
Biomatik Corporation HSD17B14 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09695
Encompass_Preferred
The HSD17B14 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human 17-beta-hydroxysteroid dehydrogenase 14 protein (1-270AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: 17-beta-hydroxysteroid dehydrogenase DHRS10, Dehydrogenase/reductase SDR family member 10, Retinal short-chain dehydrogenase/reductase retSDR3, UNQ502/PRO474, DHRS10, SDR3, HSD17B14. UniProt: Q9BPX1. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9277