Learn More
Biomatik Corporation PAIP2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09696
Encompass_Preferred
Paip2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PAIP2. The Paip2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Polyadenylate-Binding Protein-Interacting Protein 2 Protein (1-127Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PAIP2. Accession Number: Q9BPZ3. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9278