Learn More
Biomatik Corporation ADAT1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09722
Encompass_Preferred
The ADAT1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human tRNA-specific adenosine deaminase 1 protein (1-501AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: tRNA-specific adenosine deaminase 1 (EC:3.5.4.34), hADAT1, ADAT1, tRNA-specific adenosine-37 deaminase. UniProt: Q9BUB4. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9304