Learn More
Biomatik Corporation TSGA10 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09725
Encompass_Preferred
Tsga10 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TSGA10. The Tsga10 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Testis-Specific Gene 10 Protein (1-200Aa). Applications: ELISA, WB, IHC. Target Synonyms: TSGA10. Accession Number: Q9BZW7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9307