Learn More
Biomatik Corporation NAT10 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09727
Encompass_Preferred
Nat10 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NAT10. The Nat10 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Rna Cytidine Acetyltransferase Protein (909-1025Aa). Applications: ELISA, WB, IHC, IP. Target Synonyms: NAT10. Accession Number: Q9H0A0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9309