Learn More
Biomatik Corporation CLSTN2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09731
Encompass_Preferred
The CLSTN2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Calsyntenin-2 protein (473-673AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Calsyntenin-2, Alcadein-gamma, Alc-gamma, CLSTN2, CS2. UniProt: Q9H4D0. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9313