Learn More
Biomatik Corporation ATP8A2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09745
Encompass_Preferred
The ATP8A2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Phospholipid-transporting ATPase IB protein (358-570AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Phospholipid-transporting ATPase IB, ATPase class I type 8A member 2, ML-1, P4-ATPase flippase complex alpha subunit ATP8A2, ATP8A2, ATPIB. UniProt: Q9NTI2. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9327