Learn More
Biomatik Corporation FLRT1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09751
Encompass_Preferred
The FLRT1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Leucine-rich repeat transmembrane protein FLRT1 protein (21-289AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Leucine-rich repeat transmembrane protein FLRT1, Fibronectin-like domain-containing leucine-rich transmembrane protein 1, FLRT1. UniProt: Q9NZU1. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9333