Learn More
Biomatik Corporation MIOX anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09753
Encompass_Preferred
Miox Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MIOX. The Miox Polyclonal Antibody has been validated for the following applications: ELISA, WB, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Inositol Oxygenase Protein (1-285Aa). Applications: ELISA, WB, IP. Target Synonyms: MIOX. Accession Number: Q9UGB7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9335