Learn More
Biomatik Corporation FYCO1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509336
Encompass_Preferred
The FYCO1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human FYVE and coiled-coil domain-containing protein 1 protein (2-255AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: FYVE and coiled-coil domain-containing protein 1, FYCO1, ZFYVE7, Zinc finger FYVE domain-containing protein 7. UniProt: Q9BQS8. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9336 Supplier Biomatik Corporation Supplier No. CAC09754
The FYCO1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human FYVE and coiled-coil domain-containing protein 1 protein (2-255AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: FYVE and coiled-coil domain-containing protein 1, FYCO1, ZFYVE7, Zinc finger FYVE domain-containing protein 7. UniProt: Q9BQS8. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.