Learn More
Biomatik Corporation C7orf50 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09755
Encompass_Preferred
The C7orf50 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Uncharacterized protein C7orf50 protein (1-194AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-2000. Alternative Names: Uncharacterized protein C7orf50, C7orf50. UniProt: Q9BRJ6. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9337