Learn More
Biomatik Corporation SLC12A9 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09758
Encompass_Preferred
Slc12A9 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC12A9. The Slc12A9 Polyclonal Antibody has been validated for the following applications: ELISA, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Solute Carrier Family 12 Member 9 Protein (126-173Aa). Applications: ELISA, IF. Target Synonyms: SLC12A9. Accession Number: Q9BXP2. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9340