Learn More
Biomatik Corporation SH3GLB2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509350
Encompass_Preferred
Sh3Glb2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SH3GLB2. The Sh3Glb2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Endophilin-B2 Protein (1-395Aa). Applications: ELISA, WB, IHC. Target Synonyms: SH3GLB2. Accession Number: Q9NR46. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9350 Supplier Biomatik Corporation Supplier No. CAC09768
Sh3Glb2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SH3GLB2. The Sh3Glb2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Endophilin-B2 Protein (1-395Aa). Applications: ELISA, WB, IHC. Target Synonyms: SH3GLB2. Accession Number: Q9NR46. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.