Learn More
Biomatik Corporation PPP1R12C anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09775
Encompass_Preferred
Ppp1R12C Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PPP1R12C. The Ppp1R12C Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Protein Phosphatase 1 Regulatory Subunit 12C Protein (317-450Aa). Applications: ELISA, IHC, IF. Target Synonyms: PPP1R12C. Accession Number: Q9BZL4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9357