Learn More
Biomatik Corporation SLC25A32 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09779
Encompass_Preferred
Slc25A32 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC25A32. The Slc25A32 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Mitochondrial Folate Transporter; Carrier Protein (44-88Aa). Applications: ELISA, WB. Target Synonyms: SLC25A32. Accession Number: Q9H2D1. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9361