Learn More
Biomatik Corporation GHITM anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09780
Encompass_Preferred
The GHITM Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant human Growth hormone-inducible transmembrane protein (293-345AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:100-1:500, IF:1:50-1:500. Alternative Names: Growth hormone-inducible transmembrane protein, GHITM, DERP2, MICS1, TMBIM5, My021, UNQ244/PRO281, Dermal papilla-derived protein 2, Mitochondrial morphology and cristae structure 1, MICS1, Transmembrane BAX inhibitor motif-containing protein 5. UniProt: Q9H3K2. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9362