Learn More
Biomatik Corporation FAM96A anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09781
Encompass_Preferred
The FAM96A Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human MIP18 family protein FAM96A protein (1-160AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-2000. Alternative Names: MIP18 family protein FAM96A, FAM96A. UniProt: Q9H5X1. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9363